5-Aminopentan-1-ol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 5-Amino-1-pentanol
MQ200
123048-10G 123048-50G
SIAL123048-10GEA 58.1 EUR
SIAL123048-10G SIAL123048-50G
5-Aminopentan-1-ol, Sigma-Aldrich®
5-Aminopentan-1-ol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 222
Melting Pt: 36
Density: 0.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008237
CAS Number: 2508-29-4
EINECS: 219-718-2
UN: 3259
ADR: 8 III