6-Aminohexan-1-ol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 6-Amino-1-hexanol
M-Clarity™ quality level = MQ200
A56353-100G A56353-25G A56353-5G
SIALA56353-100GEA 574 EUR
SIALA56353-100G SIALA56353-25G SIALA56353-5G
6-Aminohexan-1-ol, Sigma-Aldrich®
6-Aminohexan-1-ol
Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 227
Melting Pt: 53…55 °C
MDL Number: MFCD00008241
CAS Number: 4048-33-3
EINECS: 223-748-1