1,10-Diaminodecane, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Decane-1,10-diamine

M-Clarity™ quality level = MQ100

D14204-25G D14204-5G
SIALD14204-25GEA 314 EUR
SIALD14204-25G SIALD14204-5G
1,10-Diaminodecane, Sigma-Aldrich®
1,10-Diaminodecane
Formula: C₁₀H₂₄N₂
MW: 172.31 g/mol
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00008151
CAS Number: 646-25-3
EINECS: 211-471-9
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR