Ethylenediamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1,2-Ethanediamine, 1,2-Diaminoethane
M-Clarity™ quality level = MQ200
E1521-1L E1521-250ML
SIALE1521-1LEA 118 EUR
SIALE1521-1L SIALE1521-250ML
Ethylenediamine, Sigma-Aldrich®
Ethylenediamine
Formula: C₂H₈N₂
MW: 60.1 g/mol
Boiling Pt: 117 °C (1013 hPa)
Melting Pt: 8.5 °C
Density: 0.898 g/cm³ (20 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00008204
CAS Number: 107-15-3
EINECS: 203-468-6
UN: 1604
ADR: 8,II
Merck Index: 13,03829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cbd90391008b8df01337e52ed5760383c063baa&vid=9a49702fe7150a0d3cd56afb586972447ecb6f85&prid=web&clid=SAPDAM