Ethylenediamine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1,2-Ethanediamine, 1,2-Diaminoethane
M-Clarity™ quality level = MQ200
E26266-1L
E26266-2.5L
E26266-500ML
SIALE26266-1LEA
59.8
EUR
SIALE26266-1L
SIALE26266-2.5L
SIALE26266-500ML
Ethylenediamine, Sigma-Aldrich®
Ethylenediamine
Formula:
C₂H₈N₂ MW: 60.1 g/mol Boiling Pt: 117 °C (1013 hPa) Melting Pt: 8.5 °C Density: 0.898 g/cm³ (20 °C) Flash Pt: 34 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008204 CAS Number: 107-15-3 EINECS: 203-468-6 UN: 1604 ADR: 8,II Merck Index: 13,03829 |
|
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...