1,3-Diamino-2-propanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,3-Diamino-2-hydroxypropane, 1,3-Diaminopropan-2-ol
M-Clarity™ quality level = MQ200
33262-25G-F 33262-5G-F
SIAL33262-25G-FEA 189 EUR
SIAL33262-25G-F SIAL33262-5G-F
1,3-Diamino-2-propanol, Sigma-Aldrich®
1,3-Diamino-2-propanol
Formula: C₃H₁₀N₂O
MW: 90.13 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 40…42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008142
CAS Number: 616-29-5
EINECS: 210-474-2