Dihexylamine 99+%
Supplier: Thermo Scientific
Danger
407591000 407595000
ACRO407591000EA 41.8 EUR
ACRO407591000 ACRO407595000
Dihexylamine 99+%
Dihexylamine
Formula: C₁₂H₂₇N
MW: 185.35 g/mol
Boiling Pt: 236 °C (760 mmHg)
Melting Pt: 3 °C
Density: 0.8 g/cm³
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00009521
CAS Number: 143-16-8
EINECS: 205-588-4
UN: 2927
ADR: 6.1,I
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4320 to 1.4340 (20°C, 589 nm)
GC ≥99.0 %