Dihexylamine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
131202-100ML 131202-500ML
SIAL131202-100MLEA 77.6 EUR
SIAL131202-100ML SIAL131202-500ML
Dihexylamine, Sigma-Aldrich®
Dihexylamine
Formula: C₁₂H₂₇N
MW: 185.35 g/mol
Boiling Pt: 233 °C (1013 hPa)
Melting Pt: 3 °C
Density: 0.788 g/cm³ (20 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00009521
CAS Number: 143-16-8
EINECS: 205-588-4
UN: 2927
ADR: 6.1,I