Dipropylamine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
N-Propyl-1-propanamine
M-Clarity™ quality level = MQ200
D214752-1L
D214752-500ML
SIALD214752-1LEA
63.3
EUR
SIALD214752-1L
SIALD214752-500ML
Dipropylamine, Sigma-Aldrich®
Dipropylamine
Formula:
C₆H₁₅N MW: 101.19 g/mol Boiling Pt: 109.4 Melting Pt: –63 °C Density: 0.738 g/cm³ (20 °C) Flash Pt: 7 Storage Temperature: Ambient |
MDL Number:
MFCD00009362 CAS Number: 142-84-7 EINECS: 205-565-9 UN: 2383 ADR: 3 II Merck Index: 12,03406 |