Diisopropylamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: N-Isopropyl-2-propanamine
MQ100
38290-1L-F 38290-250ML-F
SIAL38290-1L-FEA 65 EUR
SIAL38290-1L-F SIAL38290-250ML-F
Diisopropylamine, Sigma-Aldrich®
Diisopropylamine
Formula: C₆H₁₅N
MW: 101.19 g/mol
Boiling Pt: 84
Melting Pt: –61 °C
Density: 0.717 g/cm³ (20 °C)
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00008862
CAS Number: 108-18-9
EINECS: 203-558-5
UN: 1158
ADR: 3 II
Merck Index: 13,03219