N-Ethyldiisopropylamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Hünig’s base, N,N-Diisopropylethylamine, N.N’-Diisopropyl ethylamine, DIPEA
MQ200
03440-1L 03440-250ML 03440-50ML 03440-4X25ML
SIAL03440-1LEA 352 EUR
SIAL03440-1L SIAL03440-250ML SIAL03440-50ML SIAL03440-4X25ML
N-Ethyldiisopropylamine, Sigma-Aldrich®
N-Ethyldiisopropylamine
Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 127
Melting Pt: –50 °C
Density: 0.76 g/cm³ (20 °C)
Flash Pt: 6
Storage Temperature: Ambient
MDL Number: MFCD00008868
CAS Number: 7087-68-5
EINECS: 230-392-0
UN: 2733
ADR: 3 II