ESD safety shoes, lace-up, jo_Tex Low ESD, Jori
Supplier: ELTEN
12301/40 12301/41 12301/42 12301/43 12301/44 12301/45 12301/46 12301/47 12301/48
ELTN12301/40EA 65.9 EUR
ELTN12301/40 ELTN12301/41 ELTN12301/42 ELTN12301/43 ELTN12301/44 ELTN12301/45 ELTN12301/46 ELTN12301/47 ELTN12301/48
ESD safety shoes, lace-up, jo_Tex Low ESD, Jori
Shoes and Boots
Sporty ESD safety shoes made from hydrophonised microfibre, skin friendly textile lining, a full padded bellows tongue plus collar padding and a dual-density PU/PU jo_Tight sole (oil, fuel and abrasion resistant, also temperature resistant from approx. -20 to +130 °C).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full length ESD inlay sole
  • Protective steel toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifikace: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC