2-(2-Aminoethylamino)ethanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: N-(2-Hydroxyethyl)ethylenediamine, AEEA
M-Clarity™ quality level = MQ200
127582-100G 127582-1KG 127582-2KG
SIAL127582-100GEA 49.1 EUR
SIAL127582-100G SIAL127582-1KG SIAL127582-2KG
2-(2-Aminoethylamino)ethanol, Sigma-Aldrich®
2-(2-Aminoethylamino)ethanol
Formula: C₄H₁₂N₂O
MW: 104.15 g/mol
Boiling Pt: 238
Melting Pt: –28 °C
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 144
Storage Temperature: Ambient
MDL Number: MFCD00008170
CAS Number: 111-41-1
EINECS: 203-867-5