Diethanolamine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
ß,ß′-Dihydroxydiethylamine, Bis-(ß-hydroxyethyl)-amine, 2,2'-Iminodiethanol
31589-100G
31589-500G
SIAL31589-100GEA
43
EUR
SIAL31589-100G
SIAL31589-500G
Diethanolamine, Sigma-Aldrich®
Diethanolamine
Formula:
C₄H₁₁NO₂ MW: 105.14 g/mol Boiling Pt: 268.8 Melting Pt: 28 Density: 1.1 g/cm³ (20 °C) Flash Pt: 176 Storage Temperature: Ambient |
MDL Number:
MFCD00002843 CAS Number: 111-42-2 EINECS: 203-868-0 REACH: 01-2119488930-28 Merck Index: 13,03134 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...