Diethanolamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: ß,ß′-Dihydroxydiethylamine, Bis-(ß-hydroxyethyl)-amine, 2,2'-Iminodiethanol
31590-250G 31590-1KG
SIAL31590-250GEA 57.2 EUR
SIAL31590-250G SIAL31590-1KG
Diethanolamine, Sigma-Aldrich®
Diethanolamine
Formula: C₄H₁₁NO₂
Boiling Pt: 268.8 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD00002843
CAS Number: 111-42-2
EINECS: 203-868-0
REACH: 01-2119488930-28
Merck Index: 13,03134