Diethanolamine, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: ß,ß′-Dihydroxydiethylamine, Bis-(ß-hydroxyethyl)-amine, 2,2'-Iminodiethanol

M-Clarity™ quality level = MQ200

398179-500G 398179-1KG
SIAL398179-500GEA 57 EUR
SIAL398179-500G SIAL398179-1KG
Diethanolamine, Sigma-Aldrich®
Diethanolamine
Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
Boiling Pt: 268.8
Melting Pt: 28
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 176
Storage Temperature: Ambient
MDL Number: MFCD00002843
CAS Number: 111-42-2
EINECS: 203-868-0
REACH: 01-2119488930-28
Merck Index: 13,03134

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR