Diethanolamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: ß,ß′-Dihydroxydiethylamine, Bis-(ß-hydroxyethyl)-amine, 2,2'-Iminodiethanol
M-Clarity™ quality level = MQ200
D8885-500G D8885-1KG
SIALD8885-500GEA 44.8 EUR
SIALD8885-500G SIALD8885-1KG
Diethanolamine, Sigma-Aldrich®
Diethanolamine
Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
Boiling Pt: 268.8
Melting Pt: 28
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 176
Storage Temperature: Ambient
MDL Number: MFCD00002843
CAS Number: 111-42-2
EINECS: 203-868-0
REACH: 01-2119488930-28
Merck Index: 13,03134