2-Nitroethanol, Sigma-Aldrich®
Supplier: Merck
Warning
MQ100
146633-100G 146633-25G 146633-5G
SIAL146633-100GEA 509 EUR
SIAL146633-100G SIAL146633-25G SIAL146633-5G
2-Nitroethanol, Sigma-Aldrich®
2-Nitroethanol
Formula: C₂H₅NO₃
MW: 91.07 g/mol
Boiling Pt: 194
Melting Pt: –80 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007405
CAS Number: 625-48-9
EINECS: 210-895-1