Diisopropyl ether

Supplier: Roth Carl
Danger

Synonyms: Isopropyl ether

T919.1 T919.2 T919.3
ROTHT919.1EA 46.3 EUR
ROTHT919.1 ROTHT919.2 ROTHT919.3
Diisopropyl ether
Diisopropyl ether
Formula: (CH₃)₂CHOCH(CH₃)₂
MW: 102.18 g/mol
Boiling Pt: 68.5
Melting Pt: –86 °C
Density: 0.726 g/cm³ (20 °C)
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008880
CAS Number: 108-20-3
EINECS: 203-560-6
UN: 1159
ADR: 3 II
Merck Index: 13,05232
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=932b8c021b2bfa9a11b8d8812cafbafecc6f9239&vid=dea267e876203eb2c2674787701ec602da1b3cf6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR