Diethylene glycol, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 2,2'-Oxybis(ethan-1-ol), Digol, 2,2'-Oxybisethanol

M-Clarity™ quality level = MQ200

H26456-1L H26456-2.5L
SIALH26456-1LEA 36.1 EUR
SIALH26456-1L SIALH26456-2.5L
Diethylene glycol, Sigma-Aldrich®
Diethylene glycol
Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 245
Melting Pt: –10 °C
Density: 1.118 g/cm³ (20 °C)
Flash Pt: 123 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002882
CAS Number: 111-46-6
EINECS: 203-872-2
Merck Index: 13,03146
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f023bd53ecff654f6d6d398b3ee2a1bac87d56f&vid=25d8943f9c247b4203b1e1bcba59bf77dfdd9158&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR