Butyric anhydride ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98%
CAS No.: 106-31-0
Molecular Formula: C8H14O3
Formula Weight: 158.20
Flash Point: 88°(190°F)
Refractive Index: 1.4130
Odour: Rancid
Melting Point: -75°
Boiling Point: 198-199°
Beilstein Reference: 1099474
Merck Reference: 14,1594
UN No.: 2739
MDL No.: MFCD00009389
Formula:
C₈H₁₄O₃ Boiling Pt: 198…199 °C Melting Pt: –75 °C Density: 0.968 Flash Pt: 88 °C (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009389 CAS Number: 106-31-0 EINECS: 203-383-4 UN: 2739 ADR: 8,III Merck Index: 14,1594 |