Caproic anhydride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Hexanoic anhydride
M-Clarity™ quality level = MQ200
194530-250G 194530-50G
SIAL194530-250GEA 90.4 EUR
SIAL194530-250G SIAL194530-50G
Caproic anhydride, Sigma-Aldrich®
Caproic anhydride
Formula: C₁₂H₂₂O₃
MW: 214.3 g/mol
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: –40 °C
Density: 0.93 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009509
CAS Number: 2051-49-2
EINECS: 218-121-4
UN: 3265
ADR: 8,II