1-Decanol, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 1-Hydroxy decane, 1-Decyl alcohol, Capric alcohol, Decan-1-ol, Nonyl carbinol, Decyl alcohol

M-Clarity™ quality level = MQ100

150584-3KG 150584-500G
SIAL150584-3KGEA 143 EUR
SIAL150584-3KG SIAL150584-500G
1-Decanol, Sigma-Aldrich®
1-Decanol
Formula: H₃C(CH₂)₈CH₂OH
MW: 158.28 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.82634 g/cm³ (20 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00004747
CAS Number: 112-30-1
EINECS: 203-956-9
UN: 3082
ADR: 9,III
Merck Index: 13,02875

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR