1-Decanol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
1-Hydroxy decane, 1-Decyl alcohol, Capric alcohol, Decan-1-ol, Nonyl carbinol, Decyl alcohol
M-Clarity™ quality level = MQ300
W236500-1KG-K
W236500-20KG-K
W236500-8KG-K
SIALW236500-1KG-KEA
73.7
EUR
SIALW236500-1KG-K
SIALW236500-20KG-K
SIALW236500-8KG-K
1-Decanol, Sigma-Aldrich®
1-Decanol
Formula:
H₃C(CH₂)₈CH₂OH Boiling Pt: 230 °C (1013 hPa) Melting Pt: 7 °C Density: 0.82634 g/cm³ (20 °C) Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004747 CAS Number: 112-30-1 EINECS: 203-956-9 UN: 3082 ADR: 9,III Merck Index: 13,02875 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...