1-Nonanol, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIAL131210-100MLEA 51.3 EUR
SIAL131210-100ML SIAL131210-500ML
1-Nonanol, Sigma-Aldrich®
1-Nonanol
Formula: H₃C(CH₂)₇CH₂OH
MW: 144,26 g/mol
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: –7 °C
Density: 0,8419 g/cm³ (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00002990
UN: 3082
ADR: 9,III
Merck Index: 12,06774

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR