1-Nonanol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Nonan-1-ol, n-Nonanol, 1-Nonyl alcohol, Nonyl alcohol, 1-Hydroxy nonane, n-Nonyl alcohol, Octyl carbinol
M-Clarity™ quality level = MQ300
W278904-1KG-K W278904-8KG-K
SIALW278904-1KG-KEA 112 EUR
SIALW278904-1KG-K SIALW278904-8KG-K
1-Nonanol, Sigma-Aldrich®
1-Nonanol
Formula: H₃C(CH₂)₇CH₂OH
MW: 144.26 g/mol
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: –7 °C
Density: 0.8419 g/cm³ (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00002990
CAS Number: 143-08-8
EINECS: 205-583-7
UN: 3082
ADR: 9,III
Merck Index: 12,06774