1-Octadecanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
MQ200
O709-1KG O709-2KG
SIALO709-1KGEA 69 EUR
SIALO709-1KG SIALO709-2KG
1-Octadecanol, Sigma-Aldrich®
1-Octadecanol
Formula: H₃C(CH₂)₁₆CH₂OH
MW: 270.5 g/mol
Boiling Pt: 336
Melting Pt: 56…57 °C
Density: 0.805…0.815 g/cm³ (60 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002823
CAS Number: 112-92-5
EINECS: 204-017-6
Merck Index: 12,08960