3-Pentanol ≥98%, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
P8025-100ML-A P8025-25ML-A P8025-500ML-A
SIALP8025-100ML-AEA 104 EUR
SIALP8025-100ML-A SIALP8025-25ML-A SIALP8025-500ML-A
3-Pentanol ≥98%, Sigma-Aldrich®
3-Pentanol
Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 116 °C (1013 hPa)
Melting Pt: –8 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00004574
CAS Number: 584-02-1
EINECS: 209-526-7
UN: 1105
ADR: 3,III
Merck Index: 13,07197
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ec7192926b50615dfac930445c63174c6221a44&vid=a2f1fa3f1ffa1ecbfb6a5dd1d0f29da27d1d8737&prid=web&clid=SAPDAM