Aluminium isopropoxide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Aluminiumtriisopropylate, Aluminiumisopropylate, Aluminiumtriisopropoxide, Aluminium isopropoxide
MQ200
220418-100G 220418-1KG
SIAL220418-100GEA 27.4 EUR
SIAL220418-100G SIAL220418-1KG
Aluminium isopropoxide, Sigma-Aldrich®
Aluminium isopropoxide
Formula: C₉H₂₁AlO₃
MW: 204.25 g/mol
Boiling Pt: 135
Melting Pt: 119
Density: 0.9771 g/cm³ (20 °C)
MDL Number: MFCD00008870
CAS Number: 555-31-7
EINECS: 209-090-8
UN: 3181
ADR: 4.1 II
Merck Index: 13,00346