2-Mercaptoethanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Hydroxyethyl mercaptan, β-Mercaptoethanol
63689-100ML-F 63689-25ML-F
SIAL63689-100ML-FEA 90.2 EUR
SIAL63689-100ML-F SIAL63689-25ML-F
2-Mercaptoethanol, Sigma-Aldrich®
2-Mercaptoethanol
Formula: HSCH₂CH₂OH
MW: 78.14 g/mol
Boiling Pt: 157 °C (1013 hPa)
Melting Pt: –100 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00004890
CAS Number: 60-24-2
EINECS: 200-464-6
UN: 2966
ADR: 6.1,II
Merck Index: 13,05894
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc427209b84d0343a5ae47c435ddfc53df4013d2&vid=7d70fd087d948d54e7a946d3b2310e2d820d2396&prid=web&clid=SAPDAM