2-Mercaptoethanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Hydroxyethyl mercaptan, β-Mercaptoethanol
M6250-100ML M6250-1L M6250-250ML M6250-500ML
SIALM6250-100MLEA 61.8 EUR
SIALM6250-100ML SIALM6250-1L SIALM6250-250ML SIALM6250-500ML
2-Mercaptoethanol, Sigma-Aldrich®
2-Mercaptoethanol
Formula: HSCH₂CH₂OH
MW: 78.14 g/mol
Boiling Pt: 157 °C (1013 hPa)
Melting Pt: –100 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00004890
CAS Number: 60-24-2
EINECS: 200-464-6
UN: 2966
ADR: 6.1,II
Merck Index: 13,05894
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc427209b84d0343a5ae47c435ddfc53df4013d2&vid=7d70fd087d948d54e7a946d3b2310e2d820d2396&prid=web&clid=SAPDAM