1,12-Dodecanediol, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1,12-Dihydroxy dodecane

M-Clarity™ quality level = MQ100

D221309-100G D221309-25G
SIALD221309-100GEA 127 EUR
SIALD221309-100G SIALD221309-25G
1,12-Dodecanediol, Sigma-Aldrich®
1,12-Dodecanediol
Formula: C₁₂H₂₆O₂
MW: 202.34 g/mol
Boiling Pt: 313
Melting Pt: 78…79 °C
Density: 1.08 g/cm³ (20 °C)
MDL Number: MFCD00004755
CAS Number: 5675-51-4
EINECS: 227-133-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR