Glycerine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
MQ200
G6279-1L
G6279-500ML
SIALG6279-1LEA
229
EUR
SIALG6279-1L
SIALG6279-500ML
Glycerine, Sigma-Aldrich®
Glycerine
Formula:
HOCH₂CH(OH)CH₂OH MW: 92.09 g/mol Boiling Pt: 290 Melting Pt: 18.6 Density: 1.26 g/cm³ (20 °C) Flash Pt: 177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004722 CAS Number: 56-81-5 EINECS: 200-289-5 Merck Index: 13,04497 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...