Glycerine, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
MQ300
W252506-10KG-K W252506-1KG-K W252506-25KG-K
SIALW252506-10KG-KEA 295 EUR
SIALW252506-10KG-K SIALW252506-1KG-K SIALW252506-25KG-K
Glycerine, Sigma-Aldrich®
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM