Lithium dodecyl sulphate 98
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Dodecyl sulphuric acid lithium salt, Dodecyl sulphate lithium salt, Lithium lauryl sulphate, Lauryl sulphate lithium salt
A16005.06 A16005.14 A16005.22
A16005.06EA 37.3 EUR
A16005.06 A16005.14 A16005.22
Lithium dodecyl sulphate 98
Lithium dodecyl sulphate
Purity: 98% CAS No.: 2044-56-6 Molecular Formula: CH3(CH2)11OSO3Li Formula Weight: 272.33 Odour: Odourless UN No.: 1325 MDL No.: MFCD00007467
Formula: C₁₂H₂₅LiO₄S
MW: 272.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007467
CAS Number: 2044-56-6
EINECS: 218-058-2
ADR: 4.1 III