Lithium dodecyl sulphate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Dodecyl sulphuric acid lithium salt, Dodecyl sulphate lithium salt, Lithium lauryl sulphate, Lauryl sulphate lithium salt
MQ200
L5901-25G L5901-5G
SIALL5901-25GEA 378 EUR
SIALL5901-25G SIALL5901-5G
Lithium dodecyl sulphate, Sigma-Aldrich®
Lithium dodecyl sulphate
Formula: C₁₂H₂₅LiO₄S
MW: 272.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007467
CAS Number: 2044-56-6
EINECS: 218-058-2