Lithium dodecyl sulphate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Dodecyl sulphuric acid lithium salt, Dodecyl sulphate lithium salt, Lithium lauryl sulphate, Lauryl sulphate lithium salt
M-Clarity™ quality level = MQ200
L9781-25G L9781-50G L9781-5G
SIALL9781-25GEA 451 EUR
SIALL9781-25G SIALL9781-50G SIALL9781-5G
Lithium dodecyl sulphate, Sigma-Aldrich®
Lithium dodecyl sulphate
Formula: C₁₂H₂₅LiO₄S
MW: 272.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007467
CAS Number: 2044-56-6
EINECS: 218-058-2