1,6-Hexanedithiol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,6-Dimercaptohexane
M-Clarity™ quality level = MQ100
H12005-5G H12005-25G
SIALH12005-5GEA 51.8 EUR
SIALH12005-5G SIALH12005-25G
1,6-Hexanedithiol, Sigma-Aldrich®
1,6-Hexanedithiol
Formula: C₆H₁₄S₂
MW: 150.31 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: –21 °C
Density: 1.0102 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004910
CAS Number: 1191-43-1
EINECS: 214-735-1
UN: 2810
ADR: 6.1,III