1,8-Octanedithiol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
1,8-Dimercaptooctane
M-Clarity™ quality level = MQ100
O3605-1G
O3605-5G
SIALO3605-1GEA
50.2
EUR
SIALO3605-1G
SIALO3605-5G
1,8-Octanedithiol, Sigma-Aldrich®
1,8-Octanedithiol
Formula:
C₈H₁₈S₂ MW: 178.36 g/mol Boiling Pt: 270 Density: 0.9814 g/cm³ (0 °C) |
MDL Number:
MFCD00003574 CAS Number: 1191-62-4 EINECS: 214-738-8 UN: 3334 ADR: 9 |