Methanesulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Mesyl chloride
A13383.36
A13383.22
AB110764.0100
A13383.PG
A13383.36EA
51.8
EUR
A13383.36
A13383.22
ABCRAB110764.0100
A13383.PG
Methanesulphonyl chloride ≥98%
Methanesulphonyl chloride
Purity: 98%
CAS No.: 124-63-0
Molecular Formula: CH3ClO2S
Formula Weight: 114.55
Flash Point: 97°(206°F)
Refractive Index: 1.4520
Odour: Pungent
Melting Point: -33°
Boiling Point: 161°
Beilstein Reference: 506297
Merck Reference: 14,5955
UN No.: 3246
MDL No.: MFCD00007454
Formula:
CH₃ClO₂S Boiling Pt: 161 °C Melting Pt: –33 °C Density: 1.477 Flash Pt: 97 °C (206 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007454 CAS Number: 124-63-0 EINECS: 204-706-1 UN: 3246 ADR: 6.1,I Merck Index: 14,5955 |