Methanesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Mesyl chloride
M-Clarity™ quality level = MQ200
64270-1L 64270-250ML
SIAL64270-1LEA 146 EUR
SIAL64270-1L SIAL64270-250ML
Methanesulphonyl chloride, Sigma-Aldrich®
Methanesulphonyl chloride
Formula: CH₃ClO₂S
MW: 114.55 g/mol
Boiling Pt: 161 °C (1013 hPa)
Melting Pt: –32 °C
Density: 1.478 g/cm³ (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007454
CAS Number: 124-63-0
EINECS: 204-706-1
UN: 3246
ADR: 6.1,I
Merck Index: 13,05982