Sodium-1-decanesulphonate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt
M-Clarity™ quality level = MQ200
D3412-100G D3412-25G D3412-5G
SIALD3412-100GEA 850 EUR
SIALD3412-100G SIALD3412-25G SIALD3412-5G
Sodium-1-decanesulphonate, Sigma-Aldrich®
Sodium 1-decanesulphonate
Formula: C₁₀H₂₁NaO₃S
MW: 244.33 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007526
CAS Number: 13419-61-9
EINECS: 236-525-9