1-Heptanesulphonic acid sodium salt, Sigma-Aldrich®
Supplier: Merck
Synonyms: Sodium heptane-1-sulphonate, Sodium 1-heptanesulphonate
M-Clarity™ quality level = MQ100
H2766-25G H2766-100G H2766-5G
SIALH2766-25GEA 260 EUR
SIALH2766-25G SIALH2766-100G SIALH2766-5G
1-Heptanesulphonic acid sodium salt, Sigma-Aldrich®
1-Heptanesulphonic acid sodium salt
Formula: H₃C(CH₂)₅CH₂SO₃Na
MW: 202.25 g/mol
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00007543
CAS Number: 22767-50-6
EINECS: 245-210-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95c09a5691140bab7446d88919f01593e8f0b9b2&vid=7c237cde5db01d0567ff19964ca3287434747480&prid=web&clid=SAPDAM