1-Octanesulphonic acid sodium salt, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Sodium octane-1-sulphonate

M-Clarity™ quality level = MQ200

O0133-100G O0133-25G O0133-5G
SIALO0133-100GEA 496 EUR
SIALO0133-100G SIALO0133-25G SIALO0133-5G
1-Octanesulphonic acid sodium salt, Sigma-Aldrich®
1-Octanesulphonic acid sodium salt
Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=32d95cd9e321818e2f94c8261c4b4e6180ee0fc7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR