Silver methanesulphonate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
550256-25G
550256-5G
SIAL550256-25GEA
210
EUR
SIAL550256-25G
SIAL550256-5G
Silver methanesulphonate, Sigma-Aldrich®
Silver methanesulphonate
Formula:
CH₃AgO₃S Melting Pt: 263 °C |
MDL Number:
MFCD00064795 CAS Number: 2386-52-9 EINECS: 219-199-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...