n-Decane, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

457116-100ML 457116-1L 457116-2L
SIAL457116-100MLEA 125 EUR
SIAL457116-100ML SIAL457116-1L SIAL457116-2L
n-Decane, Sigma-Aldrich®
n-Decane
Formula: H₃C(CH₂)₈CH₃
Boiling Pt: 174.1 °C (1013 hPa)
Melting Pt: –30 °C
Density: 0.73 g/cm³ (20 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00008954
CAS Number: 124-18-5
EINECS: 204-686-4
UN: 2247
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e4dead3a7841a2258e8a5b99b880895cd06aa1&vid=209e0ebbe6a38426575fcf5608cf6f2b586e6d5e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR