n-Docosane, Sigma-Aldrich®
Supplier: Merck
Synonyms: N-docosane
M-Clarity™ quality level = MQ100
134457-100G 134457-25G
SIAL134457-100GEA 119 EUR
SIAL134457-100G SIAL134457-25G
n-Docosane, Sigma-Aldrich®
n-Docosane
Formula: H₃C(CH₂)₂₀CH₃
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 44 °C
Density: 0.7944 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009348
CAS Number: 629-97-0
EINECS: 211-121-5