n-Heptadecane, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
N-heptadecane
M-Clarity™ quality level = MQ100
128503-100G
128503-25G
SIAL128503-100GEA
92.1
EUR
SIAL128503-100G
SIAL128503-25G
n-Heptadecane, Sigma-Aldrich®
n-Heptadecane
Formula:
H₃C(CH₂)₁₅CH₃ MW: 240.47 g/mol Boiling Pt: 302.1 °C (1013 hPa) Melting Pt: 21.2 °C Density: 0.7783 g/cm³ (20 °C) Flash Pt: 148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009002 CAS Number: 629-78-7 EINECS: 211-108-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...