J.T.Baker® BAKERBOND Speedisk® Carboxylic Acid, SPE Columns
Supplier: Avantor
8172-04EA 0 EUR
8172-04 8172-11 8172-02 8172-06 8172-07
J.T.Baker® BAKERBOND Speedisk® Carboxylic Acid, SPE Columns
Chromatography Columns
Ultra-clean, PP columns with BAKERBOND® HPLC media are engineered to save time, minimise work and final solvent volumes and improve performance. Run times are up to nine times faster than conventional columns. The built-in column guard filter and higher analyte concentration may reduce or eliminate pre-filtration and concentration steps.
  • Run times up to nine times faster than conventional SPE columns
  • Unique laminar configuration
  • Silica based
General applications: Ionic, basic analytes.
These SPE columns can be used in weak cation exchange mode.