n-Pentadecane, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
P3406-100G P3406-500G
SIALP3406-100GEA 63.8 EUR
SIALP3406-100G SIALP3406-500G
n-Pentadecane, Sigma-Aldrich®
n-Pentadecane
Formula: H₃C(CH₂)₁₃CH₃
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 13…15 °C
Density: 0.768 g/cm³ (20 °C)
Flash Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00008990
CAS Number: 629-62-9
EINECS: 211-098-1
UN: 3295
ADR: 3,III